Learn More
Apexbio Technology LLC Losmapimod 585543-15-3 50mg
SDP
Supplier:  Apexbio Technology LLC B462050
Encompass_Preferred
Losmapimod (CAS 585543-15-3) also known as GW856553 is a selective orally bioavailable inhibitor of p38 mitogen-activated protein kinase (MAPK) It exhibits potent inhibitory activity against p38 and p38 isoforms with reported pKi values of 8 1 and 7 6 respectively By inhibiting p38 MAPK a kinase essential in inflammatory signaling pathways and cytokine production Losmapimod has demonstrated beneficial effects in preclinical and clinical models It improves nitric oxide-mediated vasodilation and reduces markers of systemic inflammation such as C-reactive protein and fibrinogen This molecule is utilized in research studies addressing inflammation-driven cardiovascular disorders hypercholesterolemia and chronic obstructive pulmonary disease (COPD)
Catalog No. 50-000-62739