Learn More
Medchemexpress LLC Lotilaner 50mg | 1369852-71-0 | 50 MG

Supplier: Medchemexpress LLC HY11656450MG
Lotilaner is an isoxazoline-class ectoparasiticide used as a research compound. It acts as a potent non-competitive antagonist of invertebrate GABACl receptors and is supplied as a high-purity solid suitable for in vitro and in vivo studies.
- Purity 99.84% (HPLC)
- Chemical formula C20H14Cl3F6N3O3S; molecular weight 596.76
- CAS number 1369852-71-0
- Available in small-mass packages including a 50 mg pack
- Powder storage: -20°C for long-term stability; in solvent: store at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.