Learn More
Medchemexpress LLC HY-116564 5mg Medchemexpress, Lotilaner CAS:1369852-71-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1165645MG
Encompass_Preferred
Medchemexpress, HY-116564 5mg Lotilaner CAS:1369852-71-0 Lotilaner is a parasiticide, acts as a potent non-competitive antagonist of insects GABACl receptors, with an IC50 of 23.84 nM for Drosophila melanogaster GABA receptor. No effect on a dog GABAA receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9431