Learn More
TARGETMOL CHEMICALS INC Loureirin D 2MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN23452MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Loureirin D is a chalcone from the herbs of Dracaena cochinchinensis. Purity 98.55%
Catalog No. 50-287-9533