Learn More
Medchemexpress LLC Lovastatin-d9 | 75330-75-5 | 413.60 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0504S11MG
Encompass_Preferred
Lovastatin-d9 is the deuterium labeled Lovastatin, a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90092