Learn More
Enzo Life Sciences Lovastatin. sodium salt (5mg). CAS: 75225-50-2
SDP
Supplier:  Enzo Life Sciences BMLG2450005
Encompass_Preferred
Active carboxylate form of Lovastatin (BML-G226). Active in whole cells as well as cell-free assays. Inhibits cholesterol/ isoprenoid biosynthesis by inhibition of HMG-CoA reductase (Ki for acid form is 1 nM) and blocks protein isoprenylation and reduces plasma cholesterol levels in humans. Causes cells to arrest early in the G1 phase. Alternative name: Mevinolin . sodium salt, 6-alpha-Methylcompactin . sodium salt. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (10mg/ml) or 100% ethanol (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0877