Learn More
ATCC LoVo; Colon Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CCL229
Encompass
LoVo is a cell lines isolated in 1971 from the large intestine of a White, 56-year-old, male with grade IV Dukes C colorectal cancer patient. LoVo cells can be used for cancer, toxicology, and immuno-oncology research.
Catalog No. 50-238-2096
 Restricted Item