Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Low-Flow Peristaltic Pump, Variable Speed, 0.14 to 4 mL/min, Manual Control; 12 VDC
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7316020
Encompass
These cost-effective peristaltic pumps provide variable flow and exceptional life performance in a compact package. Select from three different motor speeds, and three different tubing sizes to acheive flow rates from 0.03 to 4 mL/min.
Catalog No. NC9725078