Learn More
Novus Biologicals 2-40 kDa Jess or Wes Separation Module, 2 x 25 capillary cartridges
SDP
Supplier:  Novus Biologicals SMW011
Encompass
Requires Compass v3.0 or higher. Use with a Detection Module. Reagents for 50 data points includes two parts: Separation box with 2 Capillary Cartridges (25 capillaries per cartridge), 2 Jess/Wes 2-40 kDa Pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 4 and Matrix Removal Buffer), Sample Buffer, Wash Buffer (Store at RT), plus Standard Pack 5 clamshell with 2 ea. DTT, FL Standard master mix with 26 kDa System Control protein (Store at 4°C).
Catalog No. NC1366147