Learn More
Medchemexpress LLC Low molecular weight protamine | 121052-30-0 | 98.1% | 1880.18 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP510710MG
Encompass_Preferred
Low molecular weight protamine (LMWP) is a cell-penetrating peptide that demonstrates vascular endothelial growth factor (VEGF) inhibitory activity. It is utilized in cancer research due to its ability to inhibit tumor growth.
  • Cell-penetrating peptide
  • Exhibits vascular endothelial growth factor (VEGF) inhibitory activity
  • Can inhibit tumor growth
  • Used in cancer research
  • Appears as a white to off-white solid
  • Soluble in H2O: ≥ 100 mg/mL (53.19 mM)
Catalog No. 50-003-98483