Learn More
Medchemexpress LLC Low molecular weight protamine (LMWP) | 121052-30-0 | 98.07% | 1880.18 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP51075MG
Encompass_Preferred
Low molecular weight protamine is a cell-penetrating peptide with vascular endothelial growth factor (VEGF) inhibitory activity. It can inhibit tumor growth and is used in cancer research.
  • Inhibits tumor growth
  • Cell-penetrating peptide
  • Vascular endothelial growth factor inhibitory activity
  • Used in cancer research
Catalog No. 50-003-98484