Learn More
Waters Corp Waters UPLC Columns Kit, Low pH, Widest Selectivities, 4 Columns
Supplier:  Waters Corp 176001609
Encompass_Preferred
This UPLC Columns Kit is designed for method development, providing low pH compatibility and the widest selectivities for various analytical needs. It includes a selection of four columns to facilitate comprehensive method optimization.
  • Kit contains four columns.
  • Features BEH Shield RP18, BEH Phenyl, HSS C18, and HSS C18 SB chemistries.
  • BEH columns have a particle size of 1.7 μm.
  • HSS columns have a particle size of 1.8 μm.
  • Each column has dimensions of 2.1 × 50 mm.
Catalog No. 50-791-023
Add to Cart
Request a Quote