Learn More
World Precision Instrument Low Temperature Fine Tips
SDP
Supplier:  World Precision Instrument 500396
Encompass
Our Disposable Cautery Tips are designed for use with the Bovie cautery handles. Choose the tip you want based on the temperature of your cautery handle. 500390 and 500391 are the low temperature cautery handles and can burn at temperatures of 700-1200 degrees F. Use tips 500395 (1100ºF) or 500396 (1300ºF) with the low temperature handles. 500389 and 500392 are the high temperature cautery handles and can burn at temperatures of 1500- 2100 degrees F. Use tips 500393 (2000ºF) and 500394 (2200ºF) with the high temperature handles.  All the tips are individually packaged and sterile. Each box contains 10 tips. Kits are also available for high or low temperature cautery. The kits include the handle and 10 disposable cautery tips, and all the tips are fine tips.
Note : For USA order only.
Catalog No. 50-252-1446