Learn More
Instech Low-torque two channel swivel for mice; 25ga mates with tubing used in mouse Vascular Access Button™ tethers
SDP
Supplier:  Instech 375/D/25LT
Encompass
Two Channel Stainless Steel Swivels for Mice and Rats
The second fluid channel on these swivels gives you the ability to infuse and withdraw simultaneously, to infuse on one channel while monitoring on the other, or to infuse on two lines independently. Like the stainless single channel swivels, these swivels have been built to last a lifetime (follow the instructions carefully when cleaning after every use).
If you are using Instech multi-channel Vascular Access Button tethers, use the new 25ga models, 375/D/25 for rats and 375/D/25LT for mice, for an ideal fit with the 0.4mm inner diameter tubing used in these tethers.
Each swivel includes a universal swivel-to-tether clamp that connects to our button, harness, and head block tethers. Repair and replacement parts are available.
Features
  • Dual channel
  • Stainless steel body
  • Self-adjusting seals
  • Includes universal swivel-to-tether clamp
Catalog No. NC2816853