Learn More
XRF Scientific Americas Inc. PLATINUM CRUCIBLE LOW/WIDE SHAPE 100ML FROM XRF SCIENTIFIC AMERICAS
Supplier:  XRF Scientific Americas Inc. GCL100
Encompass
57x51mm with a 100 ml capacity. Lids sold separately. These 99.95% pure platinum crucibles are available in tall and wide forms, and used for Fusions, Digestion, Ash testing, Loss on Ignition (LOI), Due to their high temperature strenght, ductility, corrosion and oxidation, excellent resistance to chemical attacks by acids and fusion mixtures resistance; these platinum crucibles are critical when high temperatures & resistance to chemical attacks are required.
Catalog No. NC0056322