Learn More
Medchemexpress LLC Loxapine | 1977-10-2 | 99.9% | 327.81 | 1 ML

Supplier: Medchemexpress LLC HY1739010MM/1ML
Loxapine is an orally active dopamine inhibitor, 5-HT receptor antagonist, and a dibenzoxazepine anti-psychotic agent. It is intended for research use only.
- Orally active dopamine inhibitor
- 5-HT receptor antagonist
- Dibenzoxazepine anti-psychotic agent
- High purity of 99.93%
- Targets human 5-HT2, D4, D1, and D2 receptors
- Reduces IL-1β and IL-2 secretion in LPS-activated cultures
- Decreases serotonin (S2) receptor density in rat brain
- Available as a 10 mM solution in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.