Learn More
Medchemexpress LLC Loxapine | 1977-10-2 | 99.9% | 327.81 | 500 MG

Supplier: Medchemexpress LLC HY17390500MG
Loxapine is an orally active dopamine inhibitor, 5-HT receptor antagonist, and a dibenzoxazepine anti-psychotic agent. This compound has been shown to reduce IL-1β and IL-2 secretion by LPS-activated mixed glia cultures, and to decrease serotonin (S2) receptor density in the rat brain without elevating dopamine (D2) receptor numbers.
- Orally active dopamine inhibitor
- 5-HT receptor antagonist
- Dibenzoxazepine anti-psychotic agent
- Reduces IL-1β and IL-2 secretion in glia cultures
- Decreases serotonin (S2) receptor density in rat brain
- Targets human 5-HT2, D4, D1, D2 receptors
- Purity of 99.93%
- Molecular weight of 327.81
- Appears as a light yellow to yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.