Learn More
Apexbio Technology LLC Loxapine Succinate 27833-64-3 500mg
SDP
Supplier:  Apexbio Technology LLC B1783500
Encompass_Preferred
Loxapine Succinate (CAS 27833-64-3) is a dibenzoxazepine-based small molecule that functions as an antagonist of dopamine D2 and D4 receptors as well as serotonergic receptors By inhibiting these neurotransmitter pathways Loxapine Succinate modulates dopaminergic and serotonergic signaling in the central nervous system It is widely utilized in preclinical and mechanistic studies investigating the pharmacological modulation of dopaminergic and serotonergic systems and their roles in neuropsychiatric disorders such as schizophrenia and related behavioral phenotypes
Catalog No. 50-000-94143