Learn More
Medchemexpress LLC Loxapine (succinate) | 27833-64-3 | 98.8% | 445.90 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY17390A10MM/1ML
Encompass_Preferred
Loxapine succinate is an orally active dopamine inhibitor, 5-HT receptor antagonist, and a dibenzoxazepine anti-psychotic agent. It can also suppress bacterial efflux pump activity and inhibit intracellular multiple-antibiotic-resistant *Salmonella enterica* serovar Typhimurium in macrophages.
  • Orally active dopamine inhibitor
  • 5-HT receptor antagonist
  • Dibenzoxazepine anti-psychotic agent
  • Suppresses bacterial efflux pump activity
  • Inhibits intracellular multiple-antibiotic-resistant Salmonella enterica serovar Typhimurium in macrophages
Catalog No. 50-003-84265