Learn More
Selleck Chemical LLC Loxapine Succinate 50mg 27833-64-3
SDP
Supplier:  Selleck Chemical LLC S408650MG
Encompass_Preferred
Loxapine Succinate is a D2DR and D4DR inhibitor, serotonergic receptor antagonist and also a dibenzoxazepine anti-psychotic agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3656