Learn More
Selleck Chemical LLC Loxapine Succinate S4086-1g
SDP
Supplier:  Selleck Chemical LLC S40861G
Encompass_Preferred
Loxapine Succinate(NSC 759578) is a D2DR and D4DR inhibitor serotonergic receptor antagonist and also a dibenzoxazepine anti-psychotic agent
Catalog No. 50-310-9497