Learn More
Medchemexpress LLC Loxistatin acid | 76684-89-4 | MFCD00132882 | 99.3% | 314.38 g/mol | C15H26N2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10022710MM 1ML
Encompass_Preferred
E 64c (loxistatin acid) is an epoxide-derived inhibitor of cysteine proteases used in biochemical and cell biology research to inhibit calpain and related enzymes. Supplied as a solid and as a ready-to-use 10 mM solution in DMSO, it supports assay setup, dose-response experiments, and inhibitor profiling.
  • Epoxide-based irreversible cysteine protease inhibitor.
  • Available as solid or 10 mM solution in DMSO for experimental flexibility.
  • High purity, suitable for sensitive biochemical assays (>99%).
  • High DMSO solubility for preparation of concentrated stock solutions.
  • Useful for calpain and general cysteine protease inhibition studies.
Catalog No. 50-002-15847