Learn More
Medchemexpress LLC Loxistatin acid (E 64c) | 76684-89-4 | 99.3% | 314.38 g/mol | C15H26N2O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1002271MG
Encompass_Preferred
E 64c (Loxistatin acid) is an epoxide-derived irreversible inhibitor of cysteine proteases that primarily inhibits calpain and shows weak irreversible inhibition of cathepsin C. Supplied as a high-purity solid for research use, it is used in biochemical and cellular studies of protease activity and has been reported to exhibit entry-blocking activity against MERS-CoV.
  • Irreversible cysteine protease inhibitor targeting calpain.
  • High purity suitable for research applications.
  • Available in small solid sizes for assay flexibility.
  • Reported to exhibit entry-blocking activity against MERS-CoV.
  • Stable solid form for convenient storage and handling.
Catalog No. 50-002-15804