Learn More
Medchemexpress LLC Loxoprofen sodium | 80382-23-6 | MFCD00941425 | ≥98.0% | 268.28 g/mol | C15H17NaO3 | 10 MG

Supplier: Medchemexpress LLC HYB0578A10MG
Loxoprofen sodium is a non-steroidal, orally active anti-inflammatory agent and nonselective cyclooxygenase (COX) inhibitor used for research with analgesic and antipyretic properties. Reported activities include reduction of atherosclerosis and antitumor effects.
- Nonselective COX inhibitor with reported IC50s: COX-1 6.5 μM, COX-2 13.5 μM.
- Analgesic and antipyretic properties suitable for inflammation research.
- Molecular formula C15H17NaO3; molecular weight 268.28 g/mol.
- Typical research pack sizes include 10 mg.
- Storage: 4°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
- For research use only - not validated for clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.