Learn More
Medchemexpress LLC LP-935509 | 1454555-29-3 | 99.7% | 396.44 | C20H24N6O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117626100MG
Encompass_Preferred
LP-935509 is a potent, selective, ATP-competitive, brain-penetrant inhibitor of adaptor protein-2 associated kinase 1 (AAK1), with additional activity against BIKE and GAK. Supplied as a solid research chemical for biochemical and cellular studies.
  • Potent AAK1 inhibition with low-nanomolar IC50 (3.3 nM).
  • Also inhibits BIKE and has modest activity against GAK.
  • High reported chemical purity (99.7%).
  • Available as solid in multiple pack sizes, including 100 MG.
  • Product characterization files available: datasheet, COA, and SDS.
Catalog No. 50-002-27742