Learn More
Medchemexpress LLC Lp-PLA2-IN-1 | 1420367-28-7 | 99.2% | 468.38 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY1975710MM/1ML
Encompass_Preferred
Lp-PLA2-IN-1 is a potent Lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor. It has potential for atherosclerosis and Alzheimer's disease research. Lp-PLA2 is an enzyme involved in the hydrolysis of lipoprotein lipids or phospholipids. It travels with low-density lipoprotein (LDL) and rapidly cleaves oxidized phosphatidylcholine molecules derived from the oxidation of LDL.
  • Potent Lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor.
  • Potential for atherosclerosis research.
  • Potential for Alzheimer's disease research.
  • Involved in hydrolysis of lipoprotein lipids or phospholipids.
  • Travels with low-density lipoprotein (LDL).
  • Cleaves oxidized phosphatidylcholine molecules.
Catalog No. 50-004-01714