Learn More
Medchemexpress LLC LP44 hydrochloride | 824958-12-5 | MFCD09971090 | 98.1% | 488.13 | C27H38ClN3OS | 100 MG

Supplier: Medchemexpress LLC HY103101100MG
LP44 hydrochloride is a research compound and selective serotonin 5-HT7 receptor agonist used in in vitro and in vivo pharmacology studies. It shows high affinity for 5-HT7 (reported Ki = 0.22 nM) and is supplied as the hydrochloride salt for use in biochemical and behavioral assays.
- Selective 5-HT7 receptor agonist (Ki = 0.22 nM).
- High purity (about 98.1%).
- Hydrochloride salt, crystalline solid form.
- Soluble in DMSO (100 mg/mL); in vivo vehicle formulations provided.
- Recommended storage: solid -20°C; in solution -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.