Learn More
Cayman Chemical 1-Oleoyl Lysophosphatidic Acid (sodium salt), 22556-62-3, 1 mg
SDP
Supplier:  Cayman Chemical 622151
Encompass_Preferred
Molecular Formula C21H40O7P • Na, Purity ≥95%, Formula Weight 458.5, Synonyms: 1-O-9Z-octadecenoyl-sn-glyceryl-3-phosphoric acid, sodium salt, Oleoyl-sn-3-glycerophosphate; 1-Oleoyl lysophosphatidic acid is a species of LPA containing oleic acid at the sn-1 position. Phosphatidic acid is produced either directly through the action of PLD or through a two step process involving liberation of DAG by PLC followed by phosphorylation of DAG by diglycerol kinase.{2656} Hydrolysis of the fatty acid at the sn-2 position by PLA2 yields bioactive LPA. LPA binds to one of four different G-protein linked receptors{13001} to mediate a variety of biological responses including cell proliferation, smooth muscle contraction, platelet aggregation, neurite retraction, and cell motility.{2656} 1-Oleoyl lysophosphatidic acid is the most potent of the LPA analogs for calcium mobilization in A431 cells{2665} and for growth stimulation of a variety of cell lines.{2666}
Catalog No. NC9021739