Learn More
Cayman Chemical LPA1 Blocking Peptide
SDP
Supplier:  Cayman Chemical 100069841 EA
Encompass_Preferred
Peptide Sequence: human LPA1 amino acids 342-359 · To be used in conjunction with Cayman’s LPA1 polyclonal antibody (Catalog No. 10005280) to block protein-antibody complex formation during immunochemical analysis of LPA1.
Catalog No. 50-288-6595