Learn More
Cayman Chemical Lpd N2-3L 5mg
SDP
Supplier:  Cayman Chemical 409115 MG
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of SMLNPs encapsulating mRNA for use in vitro and in vivo; SMLNPs containing lipid N2-3L and encapsulating a luciferase reporter accumulate at the site of injection and in the draining lymph nodes of mice; lipid N2-3L-containing SMLNPs encapsulating ovalbumin mRNA and R-848 promote dendritic cell maturation and antigen presentation, reduce tumor volume, and increase survival in an MC-38-OVA murine model of colon cancer
Catalog No. 50-294-3753