Learn More
Cayman Chemical LPSSchRIchIa colI O55 2.5mg
SDP
Supplier:  Cayman Chemical 196602.5 MG
Encompass_Preferred
A form of LPS extracted from wild-type S-form E. coli serotype O55:B5; commonly used to activate TLR4 on leukocytes, eliciting inflammatory signaling in isolated cells and in vivo
Catalog No. 50-298-5025