Learn More
Biolegend LEGENDplex™ HU Inflammation Panel 2 - S/P (11-plex) w/ VbP
Supplier:  Biolegend 740884
Encompass_Preferred
LEGENDplex™ HU Inflammation Panel 2 - S/P (11-plex) w/ VbP Reactivity: Human; Apps: Multiplex - Panel; Size: 100 tests
Catalog No. 50-208-8469