Learn More
Biolegend LEGENDplex™ MU Proinflam. Chemokine Panel 2 (6-plex) w/ FP
Supplier:  Biolegend 741084
Encompass_Preferred
LEGENDplex™ MU Proinflam. Chemokine Panel 2 (6-plex) w/ FP Reactivity: Mouse; Apps: Multiplex - Panel; Size: 100 tests
Catalog No. 50-208-8638