Learn More
Medchemexpress LLC LQZ-7I | 195822-23-2 | 99.78% | 348.35 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13653850MG
Encompass_Preferred
LQZ-7I is a survivin-targeting inhibitor that inhibits survivin dimerization. It orally effectively inhibits xenograft tumor growth and induces survivin loss in tumors. LQZ-7I has improved cytotoxicity with IC50s of 3.1 μM against C4-2 cells and 4.8 μM against PC-3 cells. In vivo studies showed that LQZ-7I significantly suppresses tumor growth without notable adverse effects on mice.
  • Inhibits survivin dimerization
  • Orally effective in inhibiting xenograft tumor growth
  • Induces survivin loss in tumors
  • Cytotoxicity with IC50s of 3.1 μM against C4-2 cells
  • Cytotoxicity with IC50s of 4.8 μM against PC-3 cells
  • Suppresses tumor growth in animal models
Catalog No. 50-003-49651