Learn More
TARGETMOL CHEMICALS INC LQZ-7I 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T846925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LQZ-7I is an inhibitor of surviving-targeting. It orally effectively inhibits xenograft tumor growth and induces survivin loss in tumorspurity: 99%
Catalog No. 50-257-9070