Learn More
Aviva Systems Biology Corp PIDD1 Antibody - N-terminal region (ARP57269_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57269_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRDD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRDD. Alternate names: DKFZp434D229, MGC16925, PIDD, LRDDApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-54075