Learn More
USA Scientific Inc Tough-Spots, 1/2 inch diameter, orange, on rolls
Supplier:  USA Scientific Inc 91850503
Encompass
Tough-Spots, large, orange labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. NC9951092