Learn More
Medchemexpress LLC LRP-12 Human HEK29 50ug

Supplier: Medchemexpress LLC HYP7094650UG
Recombinant human LRP-12 protein produced in HEK293 cells with a C-terminal His tag. The purified, glycosylated protein is supplied for research use and is available in multiple quantities, including a 50 μg format; the manufacturer provides the amino acid sequence and an indicated apparent molecular weight range.
- Recombinant human LRP-12 expressed in HEK293 cells.
- C-terminal hexahistidine tag for affinity purification.
- Glycosylated protein with apparent molecular weight ~60-85 kDa.
- Supplied as purified protein in multiple sizes, including 50 μg.
- Full amino acid sequence provided for validation and assay design.
- Suitable for biochemical assays, binding studies, and antibody validation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.