Learn More
Medchemexpress LLC HY-111493 10mg Medchemexpress, LRRK2 inhibitor 1 CAS:1802525-61-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11149310MG
Encompass_Preferred
Medchemexpress, HY-111493 10mg LRRK2 inhibitor 1 CAS:1802525-61-6 LRRK2 inhibitor 1 is a potent, selective and oral LRRK2 inhibitor with an pIC50 of 6.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8886