Learn More
Medchemexpress LLC LSECtin/CLEC4G Huma 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP787785UG
Encompass_Preferred
The LSECtin/CLEC4G protein displays unique carbohydrate binding characteristics that favor mannose N-acetylglucosamine (GlcNAc) and fucose in a Ca(2 )-dependent manner as shown in vitro Notably it serves as a receptor for Japanese encephalitis virus during microbial infection highlighting its role in viral recognition and host-pathogen interactions LSECtin/CLEC4G Protein Human (Fc) is the recombinant human-derived LSECtin/CLEC4G protein expressed by E coli with N-hFc labeled tag
Catalog No. 50-001-94888