Learn More
ABclonal Technology LSM3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A131891000UL
Encompass_Preferred
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2, MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Catalog No. 50-000-97376