Learn More
Aviva Systems Biology Corp NAA38 Antibody - middle region (ARP60464_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60464_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LSMD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LSMD1. Alternate names: MGC14151, MGC74837, PFAAP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-051