Learn More
TARGETMOL CHEMICALS INC LSN2463359 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T278545MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LSN2463359 is a positive allosteric modulators of the mGlu5 receptor. purity: 99%
Catalog No. 50-258-0562