Learn More
Medchemexpress LLC LSN3318839 | 2764704-18-7 | 99.22% | C26H23Cl2N3O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14216210MM/1ML
Encompass_Preferred
LSN3318839 is an orally active positive modulator of the glucagon-like peptide-1 receptor (GLP-1R). It increases the secretion of insulin and has a blood sugar-lowering effect. For research use only.
  • Enhances potency and efficacy of GLP-1(9-36) in GLP-1R-expressed HEK293 cells
  • Increases insulin secretion in INS-1 832-3 cells with high glucose presence
  • Demonstrates hypoglycemic effect in hyperglycemic mouse models
  • Offers an improved effect when co-administered with Sitagliptin
Catalog No. 50-003-53471