Learn More
Cayman Chemical LSorbose 50g
Supplier: Cayman Chemical 2681250 G
A monosaccharide; an intermediate in the biosynthesis of L-ascorbic acid in bacteria; formed via dehydrogenation of D-sorbitol by SLDH; has commonly been used as a starting material in the commercial biosynthesis of L-ascorbic acid