Learn More
Aviva Systems Biology Corp LSS Antibody - middle region (ARP41840_P050) | 100uL
Supplier: Aviva Systems Biology Corp ARP41840_P050
This is a rabbit polyclonal antibody against LSS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LSS. Alternate names: OSCApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish