Learn More
Enzo Life Sciences Leukotriene A4 methyl ester (1mg). CAS: 73466-12-3
SDP
Supplier:  Enzo Life Sciences BMLLA0041000
Encompass_Preferred
Unstable intermediate in the biosynthesis of LTB4 and LTC4. The naturally occuring free acid is a substrate for LTA4 hydrolase and LTC4 synthase and plays a central role in transcellular leukotrieneand lipoxin biosynthesis. Mobilizes Ca2+ in human neutrophils. Supplied as the methyl ester for greater stability. Alternative name: LTA4-ME. Formulation: Oil dissolved in 2% triethylamine/hexane. Purity: ≥95% (HPLC) (remainder 11-trans isomer). Long Term Storage: -80°C.
Catalog No. 50-201-1141
Add to Cart
Request a Quote