Learn More
Cayman Chemical Leukotriene B4, 71160-24-2, 100 g
SDP
Supplier:  Cayman Chemical 20110100
Encompass_Preferred
Molecular Formula C20H32O4, Purity ≥97%, Formula Weight 336.5, Synonyms: LTB4; A key dihydroxy fatty acid derived from AA through the 5-LO pathway; promotes a number of leukocyte functions including chemotaxis and chemokinesis at subnanomolar concentrations.
Catalog No. NC9067385