Learn More
Enzo Life Sciences Leukotriene B4-dimethylamide (50µg). CAS: 83024-92-4
SDP
Supplier:  Enzo Life Sciences BMLEI1020050
Encompass_Preferred
Leukotriene B4 antagonist. Inhibits binding of [3H]-LTB4 to guinea pig lung membranes (Ki=0.13 µM). Antagonizes the degranulation response to LTB4 in rabbit PMNL (Kd=0.2 µM) acting, most likely, via receptor desensitization. Partial agonist at higher concentrations (5 µM). Formulation: Solution in ethanol. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-0545