Learn More
Cayman Chemical LthreoSphingosine d18 1
SDP
Supplier:  Cayman Chemical 1001054110 MG
Encompass_Preferred
An analog of D-erythro-sphingosine; inhibits PKC in mixed micelle assays with 50% inhibition at 2.2 mol % making it a more potent inhibitor compared to D-erythro-sphingosine or other analogs of shorter alkyl chain length
Catalog No. 50-289-1221